Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Difluorobenzoyl chloride 96%

Supplier: AMBEED, INC
Danger

A704947-1G A704947-25G A704947-5G
77104-956EA 10 USD InStock
77104-956 77104-958 77104-960
2,5-Difluorobenzoyl chloride 96%
2,5-Difluorobenzoyl chloride
Precaución: for Research Use Only


Formula: C₇H₃ClF₂O
MDL Number: MFCD00009929
CAS Number: 35730-09-7
UN: 2920
ADR: 8,3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty