Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Poziotinib ≥98%

Supplier: AMBEED, INC
Warning

A719269-1MG A719269-5MG A719269-100MG A719269-250MG
77105-024EA 6 USD InStock
77105-024 77105-026 77686-709 77686-710
Poziotinib ≥98%
Poziotinib
Precaución: for Research Use Only


Formula: C₂₃H₂₁Cl₂FN₄O₃
MDL Number: MFCD28044290
CAS Number: 1092364-38-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty