Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 1-acetyl-4-oxocyclohexanecarboxylate 95%

Supplier: AMBEED, INC
Warning

A721017-100MG A721017-250MG
77105-034EA 94 USD InStock
77105-034 77105-036
Ethyl 1-acetyl-4-oxocyclohexanecarboxylate 95%
Ethyl 1-acetyl-4-oxocyclohexanecarboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₆O₄
MDL Number: MFCD26394906
CAS Number: 72653-21-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty