Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Phthaloyl-L-phenylalanine 98%

Supplier: AMBEED, INC
Warning

A724312-100G A724312-1G A724312-25G A724312-5G
77105-050EA 73 USD InStock
77105-050 77105-052 77105-054 77105-056
N-Phthaloyl-L-phenylalanine 98%
N-Phthaloyl-L-phenylalanine
Precaución: for Research Use Only


Formula: C₁₇H₁₃NO₄
MW: 295.29 g/mol
MDL Number: MFCD00069738
CAS Number: 5123-55-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty