Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Methoxy-2,3,4,5-tetrahydro-1H-benzo[b]azepine 95%

Supplier: AMBEED, INC
Warning

A748507-100MG A748507-1G A748507-250MG
77105-236EA 70 USD InStock
77105-236 77105-238 77105-240
8-Methoxy-2,3,4,5-tetrahydro-1H-benzo[b]azepine 95%
8-Methoxy-2,3,4,5-tetrahydro-1H-benzo[b]azepine
Precaución: for Research Use Only


Formula: C₁₁H₁₅NO
MDL Number: MFCD09026882
CAS Number: 17422-43-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty