Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichloroanisole 98%

Supplier: AMBEED, INC
Warning

A754930-100G A754930-1G A754930-25G A754930-5G
77105-278EA 112 USD InStock
77105-278 77105-280 77105-282 77105-284
3,5-Dichloroanisole 98%
3,5-Dichloroanisole
Precaución: for Research Use Only


Formula: C₇H₆Cl₂O
MDL Number: MFCD00000589
CAS Number: 33719-74-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty