Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethylene glycol dibenzoate 95%

Supplier: AMBEED, INC
Danger

A767871-1G A767871-250MG
77105-372EA 23 USD InStock
77105-372 77105-374
Ethylene glycol dibenzoate 95%
Ethylene glycol dibenzoate
Precaución: for Research Use Only


Formula: C₁₆H₁₄O₄
MDL Number: MFCD00059685
CAS Number: 94-49-5
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty