Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-acetylbenzoate 90%

Supplier: AMBEED, INC
Warning

A771438-100MG A771438-1G A771438-250MG
77105-406EA 20 USD InStock
77105-406 77105-408 77105-410
Methyl 2-acetylbenzoate 90%
Methyl 2-acetylbenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₃
MW: 178.18 g/mol
MDL Number: MFCD20485857
CAS Number: 1077-79-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty