Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-[3-(Dimethylamino)allylidene]-N-methylmethanaminium Hexafluorophosphate 98%

Supplier: AMBEED, INC
Warning

A778211-1G A778211-5G
77105-420EA 35 USD InStock
77105-420 77105-422
N-[3-(Dimethylamino)allylidene]-N-methylmethanaminium Hexafluorophosphate 98%
N-[3-(Dimethylamino)allylidene]-N-methylmethanaminium Hexafluorophosphate
Precaución: for Research Use Only


Formula: C₇H₁₅F₆N₂P
MDL Number: MFCD09788576
CAS Number: 188826-61-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty