Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,5,5-Tetramethyl-2-(2-methyl-4-(methylsulfonyl)phenyl)-1,3,2-dioxaborolane 97%

Supplier: AMBEED, INC
Warning

A789724-100MG A789724-1G A789724-250MG
77105-514EA 39 USD InStock
77105-514 77105-516 77105-518
4,4,5,5-Tetramethyl-2-(2-methyl-4-(methylsulfonyl)phenyl)-1,3,2-dioxaborolane 97%
4,4,5,5-Tetramethyl-2-(2-methyl-4-(methylsulfonyl)phenyl)-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₄H₂₁BO₄S
MDL Number: MFCD18732808
CAS Number: 1011459-26-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty