Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Bromo-8-methoxyquinoline 95%

Supplier: AMBEED, INC
Warning

A793550-100MG A793550-1G A793550-250MG A793550-50MG
77105-526EA 17 USD InStock
77105-526 77105-528 77105-530 77105-532
7-Bromo-8-methoxyquinoline 95%
7-Bromo-8-methoxyquinoline
Precaución: for Research Use Only


Formula: C₁₀H₈BrNO
MDL Number: MFCD18452260
CAS Number: 36748-99-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty