Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hyperosid 97%

Supplier: AMBEED, INC
Warning

A795483-1MG A795483-100MG A795483-250MG A795483-50MG
77105-556EA 10 USD InStock
77105-556 77686-818 77686-819 77686-820
Hyperosid 97%
Hyperosid
Precaución: for Research Use Only


Formula: C₂₁H₂₀O₁₂
MDL Number: MFCD00016933
CAS Number: 482-36-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty