Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-(Benzyloxy)benzyl)hydrazine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A805578-100MG A805578-1G A805578-250MG
77105-614EA 8 USD InStock
77105-614 77105-616 77105-618
(3-(Benzyloxy)benzyl)hydrazine hydrochloride 97%
(3-(Benzyloxy)benzyl)hydrazine hydrochloride
Precaución: for Research Use Only


Formula: C₁₄H₁₇ClN₂O
MW: 264.75 g/mol
MDL Number: MFCD09025320
CAS Number: 40051-69-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty