Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Acetyl-3,5-dimethyl-1-adamantanamine 97%

Supplier: AMBEED, INC
Warning

A820302-1G A820302-250MG A820302-5G A820302-25G
77105-730EA 39 USD InStock
77105-730 77105-732 77105-734 77686-845
N-Acetyl-3,5-dimethyl-1-adamantanamine 97%
N-Acetyl-3,5-dimethyl-1-adamantanamine
Precaución: for Research Use Only


Formula: C₁₄H₂₃NO
MDL Number: MFCD06656139
CAS Number: 19982-07-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty