Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dihydronaphthalene 98%

Supplier: AMBEED, INC
Warning

A840222-100G A840222-1G A840222-25G A840222-5G
77105-862EA 384 USD InStock
77105-862 77105-864 77105-866 77105-868
1,2-Dihydronaphthalene 98%
1,2-Dihydronaphthalene
Precaución: for Research Use Only


Formula: C₁₀H₁₀
MDL Number: MFCD00001672
CAS Number: 447-53-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty