Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-hydroxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A841137-1G A841137-250MG A841137-5G A841137-25G
77105-870EA 25 USD InStock
77105-870 77105-872 77105-874 77686-872
4-Chloro-2-hydroxyphenylboronic acid 98%
4-Chloro-2-hydroxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₆BClO₃
MW: 172.37 g/mol
MDL Number: MFCD18383546
CAS Number: 1238196-66-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty