Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,7-Dihydroxyquinazoline 95%

Supplier: AMBEED, INC
Warning

A844059-100MG A844059-250MG
77105-880EA 19 USD InStock
77105-880 77105-882
4,7-Dihydroxyquinazoline 95%
4,7-Dihydroxyquinazoline
Precaución: for Research Use Only


Formula: C₈H₆N₂O₂
MDL Number: MFCD01823180
CAS Number: 16064-25-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty