Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Difluorophenylacetyl chloride 95%

Supplier: AMBEED, INC
Danger

A847652-100MG A847652-1G A847652-250MG A847652-5G
77105-900EA 36 USD InStock
77105-900 77105-902 77105-904 77105-906
2,5-Difluorophenylacetyl chloride 95%
2,5-Difluorophenylacetyl chloride
Precaución: for Research Use Only


Formula: C₈H₅ClF₂O
MW: 190.575 g/mol
MDL Number: MFCD03424719
CAS Number: 157033-23-3
UN: 1760
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty