Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-2-naphthonitrile 95%

Supplier: AMBEED, INC
Warning

A847790-100MG A847790-1G A847790-250MG
77105-908EA 27 USD InStock
77105-908 77105-910 77105-912
3-Hydroxy-2-naphthonitrile 95%
3-Hydroxy-2-naphthonitrile
Precaución: for Research Use Only


Formula: C₁₁H₇NO
MW: 169.179 g/mol
MDL Number: MFCD17012306
CAS Number: 52449-77-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty