Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 98%

Supplier: AMBEED, INC
Warning

A852763-100MG A852763-1G A852763-250MG A852763-5G
77105-932EA 39 USD InStock
77105-932 77105-934 77105-936 77105-938
Methyl 3-cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 98%
Methyl 3-cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Precaución: for Research Use Only


Formula: C₁₅H₁₈BNO₄
MDL Number: MFCD18373989
CAS Number: 1352413-06-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty