Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,4R)-(-)-Ethyl-4-hydroxy-2-pyrrolidincarboxylate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A854707-100G A854707-25G A854707-500G
77105-944EA 70 USD InStock
77105-944 77105-946 77105-948
(2S,4R)-(-)-Ethyl-4-hydroxy-2-pyrrolidincarboxylate hydrochloride 98%
(2S,4R)-(-)-Ethyl-4-hydroxy-2-pyrrolidincarboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₃NO₃·HCl
MDL Number: MFCD00040589
CAS Number: 33996-30-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty