Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(4-methoxycarbonylphenyl) terephthalate 98+%

Supplier: AMBEED, INC
Warning

A857115-100G A857115-1G A857115-25G A857115-5G
77105-956EA 220 USD InStock
77105-956 77105-958 77105-960 77105-962
Bis(4-methoxycarbonylphenyl) terephthalate 98+%
Bis(4-methoxycarbonylphenyl) terephthalate
Precaución: for Research Use Only


Formula: C₂₄H₁₈O₈
MDL Number: MFCD00145360
CAS Number: 99328-23-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty