Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α'-Bromo-α,α,α-trifluoro-m-xylene 98%

Supplier: AMBEED, INC
Danger

A869946-10G A869946-1G A869946-25G A869946-5G
77106-004EA 20 USD InStock
77106-004 77106-006 77106-008 77106-010
α'-Bromo-α,α,α-trifluoro-m-xylene 98%
α'-Bromo-α,α,α-trifluoro-m-xylene
Precaución: for Research Use Only


Formula: C₈H₆BrF₃
MW: 239.03
Boiling Pt: 50
Density: 1.56
MDL Number: MFCD00000395
CAS Number: 402-23-3
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty