Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloromethyl isopropyl carbonate 98%

Supplier: AMBEED, INC
Danger

A882233-25G A882233-5G
77106-078EA 6 USD InStock
77106-078 77106-080
Chloromethyl isopropyl carbonate 98%
Chloromethyl isopropyl carbonate
Precaución: for Research Use Only


Formula: C₅H₉ClO₃
MDL Number: MFCD07375443
CAS Number: 35180-01-9
UN: 2924
ADR: 3,8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty