Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

XEN445 98%

Supplier: AMBEED, INC
Warning

A885460-10MG A885460-1MG A885460-25MG A885460-50MG A885460-5MG
77106-098EA 31 USD InStock
77106-098 77106-100 77106-102 77106-104 77106-106
XEN445 98%
XEN445
Precaución: for Research Use Only


Formula: C₁₈H₁₇F₃N₂O₃
MW: 366.33 g/mol
MDL Number: MFCD28166367
CAS Number: 1515856-92-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty