Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Methoxypyrimidin-5-yl)methanol 95%

Supplier: AMBEED, INC
Warning

A892528-100MG A892528-1G A892528-250MG
77106-146EA 20 USD InStock
77106-146 77106-148 77106-150
(2-Methoxypyrimidin-5-yl)methanol 95%
(2-Methoxypyrimidin-5-yl)methanol
Precaución: for Research Use Only


Formula: C₆H₈N₂O₂
MDL Number: MFCD11840201
CAS Number: 1158735-09-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty