Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-4-fluorobenzoate 95%

Supplier: AMBEED, INC
Warning

A904051-25G A904051-5G A904051-100G
77106-188EA 26 USD InStock
77106-188 77106-190 77686-944
Sodium-4-fluorobenzoate 95%
Sodium-4-fluorobenzoate
Precaución: for Research Use Only


Formula: C₇H₄FO₂Na
MDL Number: MFCD01661384
CAS Number: 499-90-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty