Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-N-[1-(5,6,7,8-Tetrahydronaphthalen-1-yl)ethyl]-3-[3-(trifluoromethyl)phenyl]-1-propylamine 98%

Supplier: AMBEED, INC
Warning

A915884-100MG A915884-1G A915884-250MG A915884-50MG
77106-222EA 64 USD InStock
77106-222 77106-224 77106-226 77106-228
(R)-N-[1-(5,6,7,8-Tetrahydronaphthalen-1-yl)ethyl]-3-[3-(trifluoromethyl)phenyl]-1-propylamine 98%
(R)-N-[1-(5,6,7,8-Tetrahydronaphthalen-1-yl)ethyl]-3-[3-(trifluoromethyl)phenyl]-1-propylamine
Precaución: for Research Use Only


Formula: C₂₂H₂₆F₃N
MDL Number: MFCD28167920
CAS Number: 1229224-93-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty