Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxycarbonyl-5-trifluoromethoxylphenylboronic acid, pinacol ester 95%

Supplier: AMBEED, INC
Warning

A927362-100MG A927362-250MG
77106-262EA 40 USD InStock
77106-262 77106-264
3-Methoxycarbonyl-5-trifluoromethoxylphenylboronic acid, pinacol ester 95%
3-Methoxycarbonyl-5-trifluoromethoxylphenylboronic acid, pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₁₈BF₃O₅
MDL Number: MFCD12026096
CAS Number: 1150561-63-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty