Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Monomethyl 3-methylglutarate 98%

Supplier: AMBEED, INC
Warning

A945144-100G A945144-25G A945144-5G A945144-10G
77106-328EA 356 USD InStock
77106-328 77106-330 77106-332 77686-974
Monomethyl 3-methylglutarate 98%
Monomethyl 3-methylglutarate
Precaución: for Research Use Only


Formula: C₇H₁₂O₄
MDL Number: MFCD01631203
CAS Number: 27151-65-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty