Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

A66 98%

Supplier: AMBEED, INC
Warning

A102947-10MG A102947-1MG A102947-50MG A102947-5MG
77107-298EA 117.42 USD InStock
77107-298 77107-300 77107-302 77107-304
A66 98%
A66
Precaución: for Research Use Only


Formula: C₁₇H₂₃N₅O₂S₂
MDL Number: MFCD22378485
CAS Number: 1166227-08-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty