Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PND-1186 98%

Supplier: AMBEED, INC
Warning

A103079-10MG A103079-1MG A103079-250MG A103079-2MG A103079-50MG A103079-5MG
77107-320EA 29.86 USD InStock
77107-320 77107-322 77107-324 77107-326 77107-328 77107-330
PND-1186 98%
PND-1186
Precaución: for Research Use Only


Formula: C₂₅H₂₆F₃N₅O₃
MDL Number: MFCD28125506
CAS Number: 1061353-68-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty