Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-3-piperidinemethanol 97%

Supplier: AMBEED, INC
Warning

A103093-100G A103093-10G A103093-25G A103093-500G A103093-5G
77107-332EA 112.27 USD InStock
77107-332 77107-334 77107-336 77107-338 77107-340
1-Methyl-3-piperidinemethanol 97%
1-Methyl-3-piperidinemethanol
Precaución: for Research Use Only


Formula: C₇H₁₅NO
MDL Number: MFCD00006497
CAS Number: 7583-53-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty