Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-3-formylpyridine 98%

Supplier: AMBEED, INC
Warning

A103855-100MG A103855-1G A103855-250MG A103855-25G A103855-5G
77107-380EA 5.15 USD InStock
77107-380 77107-382 77107-384 77107-386 77107-388
4-Amino-3-formylpyridine 98%
4-Amino-3-formylpyridine
Precaución: For research use only.


Formula: C₆H₆N₂O
MW: 122.13
MDL Number: MFCD04038972
CAS Number: 42373-30-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty