Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Chlorophenyl)-4-phenyl-4,5-dihydro-1H-pyrazole 95%

Supplier: AMBEED, INC
Warning

A104848-100G A104848-100MG A104848-1G A104848-250MG A104848-5G
77107-468EA 3149.74 USD InStock
77107-468 77107-470 77107-472 77107-474 77107-476
3-(4-Chlorophenyl)-4-phenyl-4,5-dihydro-1H-pyrazole 95%
3-(4-Chlorophenyl)-4-phenyl-4,5-dihydro-1H-pyrazole
Precaución: for Research Use Only


Formula: C₁₅H₁₃ClN₂
MDL Number: MFCD07776855
CAS Number: 59074-26-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty