Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

p,α,α,α-Tetrafluoroanisole 98%

Supplier: AMBEED, INC
Warning

A104891-100G A104891-10G A104891-25G A104891-5G
77107-488EA 77.24 USD InStock
77107-488 77107-490 77107-492 77107-494
p,α,α,α-Tetrafluoroanisole 98%
p,α,α,α-Tetrafluoroanisole
Precaución: for Research Use Only


Formula: C₇H₄F₄O
MW: 180.1 g/mol
MDL Number: MFCD00040835
CAS Number: 352-67-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty