Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-fluorotoluene 96%

Supplier: AMBEED, INC
Warning

A105180-100G A105180-10G A105180-1G A105180-25G A105180-5G
77107-508EA 92.71 USD InStock
77107-508 77107-510 77107-512 77107-514 77107-516
2-Bromo-6-fluorotoluene 96%
2-Bromo-6-fluorotoluene
Precaución: for Research Use Only


Formula: C₇H₆BrF
MW: 189.03
MDL Number: MFCD00051438
CAS Number: 1422-54-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty