Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Chlorophenoxy)piperidine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A105211-100MG A105211-1G A105211-250MG A105211-5G
77107-518EA 25.75 USD InStock
77107-518 77107-520 77107-522 77107-524
4-(2-Chlorophenoxy)piperidine hydrochloride 95%
4-(2-Chlorophenoxy)piperidine hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₅Cl₂NO
MW: 248.15 g/mol
MDL Number: MFCD02684107
CAS Number: 849107-20-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty