Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Pyridylmethanol 97%

Supplier: AMBEED, INC
Warning

A105479-100G A105479-25G A105479-500G A105479-5G A105479-10G
77107-546EA 41.21 USD InStock
77107-546 77107-548 77107-550 77107-552 77684-956
2-Pyridylmethanol 97%
2-Pyridylmethanol
Precaución: for Research Use Only


Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 223
Melting Pt: -6
MDL Number: MFCD00006348
CAS Number: 586-98-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty