Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Mercapto-5-(trifluoromethyl)pyridine 97%

Supplier: AMBEED, INC
Warning

A105818-100G A105818-10G A105818-25G A105818-5G
77107-590EA 76.22 USD InStock
77107-590 77107-592 77107-594 77107-596 77107-598
2-Mercapto-5-(trifluoromethyl)pyridine 97%
2-Mercapto-5-(trifluoromethyl)pyridine
Precaución: for Research Use Only


Formula: C₆H₄F₃NS
MDL Number: MFCD00128893
CAS Number: 76041-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty