Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,5-Trichlorobenzoic acid 95%

Supplier: AMBEED, INC
Warning

A108403-10G A108403-1G A108403-250MG A108403-25G A108403-5G
77107-814EA 5.31 USD InStock
77107-814 77107-816 77107-818 77107-820 77107-822 77107-824
2,4,5-Trichlorobenzoic acid 95%
2,4,5-Trichlorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃Cl₃O₂
MDL Number: MFCD01662044
CAS Number: 50-82-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty