Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-morpholinoaniline 98%

Supplier: AMBEED, INC
Warning

A108435-10G A108435-25G A108435-500G A108435-5G A108435-100G
77107-826EA 7.21 USD InStock
77107-826 77107-828 77107-830 77107-832 77684-972
3-Fluoro-4-morpholinoaniline 98%
3-Fluoro-4-morpholinoaniline
Precaución: for Research Use Only


Formula: C10H13FN2O
MDL Number: MFCD02571270
CAS Number: 93246-53-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty