Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SB-505124 98%

Supplier: AMBEED, INC
Warning

A114347-10MG A114347-1MG A114347-5MG
77108-196EA 80.34 USD InStock
77108-196 77108-198 77108-200
SB-505124 98%
SB-505124
Precaución: for Research Use Only


Formula: C₂₀H₂₁N₃O₂
MW: 335.4 g/mol
MDL Number: MFCD11045901
CAS Number: 694433-59-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty