Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl (2-amino-4-thiazolyl)acetate 97%

Supplier: AMBEED, INC
Warning

A116931-10G A116931-25G A116931-5G
77108-282EA 6.18 USD InStock
77108-282 77108-284 77108-286
Ethyl (2-amino-4-thiazolyl)acetate 97%
Ethyl (2-amino-4-thiazolyl)acetate
Precaución: for Research Use Only


Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
MDL Number: MFCD00005330
CAS Number: 53266-94-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty