Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TCS PIM-1 1 ≥98%

Supplier: AMBEED, INC
Warning

A118376-10MG A118376-1MG A118376-50MG A118376-5MG
77108-400EA 42.23 USD InStock
77108-400 77108-402 77108-404 77108-406
TCS PIM-1 1 ≥98%
TCS PIM-1 1
Precaución: for Research Use Only


Formula: C₁₈H₁₁BrN₂O₂
MDL Number: MFCD00276275
CAS Number: 491871-58-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty