Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridine-N-oxide 97%

Supplier: AMBEED, INC
Warning

A119987-100G A119987-25G A119987-500G A119987-5G
77108-514EA 60.77 USD InStock
77108-514 77108-516 77108-518 77108-520
Pyridine-N-oxide 97%
Pyridine-N-oxide
Precaución: for Research Use Only


Formula: C₅H₅NO
MDL Number: MFCD00006194
CAS Number: 694-59-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty