Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-5-(hydroxymethyl)phenol 97%

Supplier: AMBEED, INC
Warning

A124668-250MG A124668-5G
77108-774EA 680.04 USD InStock
77108-774 77108-776 77108-778
2-Fluoro-5-(hydroxymethyl)phenol 97%
2-Fluoro-5-(hydroxymethyl)phenol
Precaución: for Research Use Only


Formula: C₇H₇FO₂
MDL Number: MFCD12031832
CAS Number: 934241-78-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty