Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-fluorobenzoic acid 95%

Supplier: AMBEED, INC
Warning

A127468-10G A127468-25G A127468-5G
77108-988EA 11.33 USD InStock
77108-988 77108-990 77108-992 77108-994
2-Chloro-5-fluorobenzoic acid 95%
2-Chloro-5-fluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄ClFO₂
MDL Number: MFCD00156967
CAS Number: 2252-50-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty