Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Bis(morpholinomethyl)benzene-1,4-diol 95%

Supplier: AMBEED, INC
Warning

A129848-100MG A129848-50MG
77109-158EA 147.28 USD InStock
77109-158 77109-160 77109-162 77109-164 77109-166 77109-168
2,5-Bis(morpholinomethyl)benzene-1,4-diol 95%
2,5-Bis(morpholinomethyl)benzene-1,4-diol
Precaución: for Research Use Only


Formula: C₁₆H₂₄N₂O₄
MDL Number: MFCD00087356
CAS Number: 6616-56-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty