Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(2,5-Dichlorophenyl)-1,3,5-triazine-2,4-diamine 98%

Supplier: AMBEED, INC
Warning

A130508-1G A130508-1MG A130508-250MG A130508-25MG A130508-5G
77109-210EA 78.28 USD InStock
77109-210 77109-212 77109-214 77109-216 77109-218
6-(2,5-Dichlorophenyl)-1,3,5-triazine-2,4-diamine 98%
6-(2,5-Dichlorophenyl)-1,3,5-triazine-2,4-diamine
Precaución: for Research Use Only


Formula: C₉H₇Cl₂N₅
MW: 256.09 g/mol
MDL Number: MFCD00866871
CAS Number: 57381-26-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty